Learn More
Hettich™ Adapter for Carrier no. 4780
Product Code. 10288104
Change view
Click to view available options
Capacity:
14 x 12 mL
2 x 100 mL
24 x 1.5 to 2 mL Tubes
250 mL
35 x 1.1 to 5 mL Tubes
4 x 50 mL
400 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
Product Code. Capacity unitSize
10288104 7 x 25 to 30 mL Tubes Each
10156344 2 x 100 mL Each
10639453 400 mL Each
10785294 35 x 1.1 to 5 mL Tubes Each
10685515 4 x 50 mL Each
10515895 250 mL Each
10699443 24 x 1.5 to 2 mL Tubes Each
10396353 14 x 12 mL Each
8 options
Product Code. 10288104
Brand: Hettich™ 4764
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 4780
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Round Bottom, Conical, Skirted
Capacity 7 x 25 to 30 mL Tubes
Max. Tube Diameter 26 mm
Max. Tube Length 110 mm
For Use With (Equipment) Carrier no. 4780
Vessels per Adapter 7