Learn More
Hettich™ Adapter for Carrier no. 4780
Description
- Adapter for Carrier no. 4780
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom |
| Capacity | 2 x 100 mL |
| Max. Tube Diameter | 44 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Carrier no. 4780 |
| Vessels per Adapter | 2 |