Learn More
Hettich™ Adapter for Carrier no. 4275
Click to view available options
Capacity:
100 mL Tube
Unit Size:
Each
Description
- Adapter for Carrier no. 4275
- For use with ASTM tubes
- Compatible with Hettich Rotors no. 4274 and 4475
Specifications
Specifications
| Type | Adapter |
| Tube Type | ASTM |
| Capacity | 100 mL Tube |
| Max. Tube Diameter | 44 mm |
| Max. Tube Length | 168 mm |
| For Use With (Equipment) | Hettich Carrier no. 4275 |
| Vessels per Adapter | 1 |
| No. of Places | 1 |