Learn More
Hettich™ Adapter for Carrier no. 1427, for Conical Tubes
Product Code. 10001932
missing translation for 'orderingAttributeHoverText'
Capacity:
1 x 50 mL Tube
missing translation for 'unitSize'
Each
Product Code. 10001932
missing translation for 'mfr': Hettich™ 5276
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 1427
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Conical
Capacity 1 x 50 mL Tube
Max. Tube Diameter 29 mm
Max. Tube Length 115 mm
For Use With (Equipment) Carrier no. 1427
Vessels per Adapter 1