Learn More
Hettich™ Adapter for Carrier no. 1427
Click to view available options
Capacity:
12 x 2.7 to 6 mL Tubes
12 x 4.5 to 7 mL Tubes
15 mL
2 x 25 mL
50 mL
6 x 8.5 to 15 mL Tubes
6 x 9 mL
9 x 1.5 to 2 mL Tubes
Unit Size:
Each
Description
- Adapter for Carrier no. 1427
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Blood/Urine, Round Bottom |
| Capacity | 12 x 2.7 to 6 mL Tubes |
| Max. Tube Diameter | 12 mm |
| Max. Tube Length | 82 mm |
| For Use With (Equipment) | Carrier no. 1427 |
| Vessels per Adapter | 12 |
Product Content Correction
Product Title
Hettich™ Adapter for Carrier no. 1427 >