Learn More
Hettich™ Adapter for Carrier no. 1427
Product Code. 10738883
Change view
Click to view available options
Capacity:
12 x 2.7 to 6 mL Tubes
12 x 4.5 to 7 mL Tubes
15 mL
2 x 25 mL
50 mL
6 x 8.5 to 15 mL Tubes
6 x 9 mL
9 x 1.5 to 2 mL Tubes
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10738883 12 x 2.7 to 6 mL Tubes Each
12003924 12 x 4.5 to 7 mL Tubes Each
10316562 6 x 8.5 to 15 mL Tubes Each
10633333 2 x 25 mL Each
10744084 50 mL Each
10514473 6 x 9 mL Each
10021942 9 x 1.5 to 2 mL Tubes Each
12043924 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10738883 Supplier Hettich™ Supplier No. 5229
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 1427
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Blood/Urine, Round Bottom
Capacity 12 x 2.7 to 6 mL Tubes
Max. Tube Diameter 12 mm
Max. Tube Length 82 mm
For Use With (Equipment) Carrier no. 1427
Vessels per Adapter 12
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.