Learn More
Hettich™ Adapter for Carrier 5051
Description
- Adapter for Carrier no. 5051
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom, Conical |
| Capacity | 12 x 4 to 7 mL Tubes |
| Max. Tube Diameter | 13 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Carrier 5051 |
| Vessels per Adapter | 12 |