Learn More
Hettich™ Adapter for Carrier 5051
Product Code. 10265442
Change view
Click to view available options
Capacity:
11 x 8.5 to 11 mL Tubes
12 x 4 to 7 mL Tubes
2 x 50 mL
7 x 15 mL
Unit Size:
Each
Product Code. Capacity unitSize
10265442 2 x 50 mL Each
10590723 7 x 15 mL Each
10783323 11 x 8.5 to 11 mL Tubes Each
12003934 12 x 4 to 7 mL Tubes Each
4 options
Product Code. 10265442
Brand: Hettich™ 5259
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 5051
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Conical
Capacity 2 x 50 mL
Max. Tube Diameter 29 mm
Max. Tube Length 115 mm
For Use With (Equipment) Carrier 5051
Vessels per Adapter 2