Learn More
WTW™ Adapter Cable
For connection of 2 electrode conductivity measuring cells and temperature probes.
Brand: WTW™ 301429
Additional Details : Weight : 1.00000kg
Description
- Accessory for special conductivity measuring cells
- Aids in water analytics applications
- For Temperature sensor models TFK 325 or TFK 150
Specifications
Adapter Cable | |
Water analytics | |
2-electrode Conductivity Cells and Temperature Sensors TFK 325 or TFK 150 |