Learn More
Motic™ Adapter, Brightfield, LM Plan
Description
- Accommodates brightfield LED.
- For LM Plan objective
- Leverage advantages of brightfield microscopy:
- Optics do not change observed structure color.
- Optics do not change stain colors.
- Generally less expensive than phase contrast optics.
- Requires fewer adjustments for specimen observation.
Specifications
Specifications
| Type | Bright Field |
| For Use With (Equipment) | LM Plan Objective, AE2000MET Microscopes |