Learn More
Hettich™ Adapter for Blood and Urine Tubes, for EBA 200 Benchtop Centrifuges
Description
- Adapter for blood/urine tubes 13 x 75mm (dia. x L)
- For up to 8 tubes
- RPM/RCF: less than or equal to 8, 000/6, 153
- Frame with all-metal containment
Specifications
Specifications
| Type | Adapter for Blood/Urine Tubes |
| For Use With (Equipment) | EBA 200 Benchtop Centrifuge |
| Capacity | 8 Tubes |
| Description | Frame with all-metal containment |
| Tube Size | 13 x 75 mm |