Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6874
Additional Details : Weight : 0.08000kg
Description
- Adapter
Specifications
Thread Size: M24 x 1.5 female to 3/4 NPT female | |
Thermostats | |
Adapter |