Learn More
Hettich™ Adapter for Angle Rotors
Click to view available options
Capacity:
1 x 1.5 to 2 mL
Unit Size:
Pack of 6
Description
- For use with a wide range of Hettich rotors
- Holds one microliter tube 11 x 38mm; 1.5 to 2mL
Specifications
Specifications
| Type | Adapter |
| Tube Type | Microtube |
| Capacity | 1 x 1.5 to 2 mL |
| Max. Tube Diameter | 11 mm |
| Max. Tube Length | 38 mm |
| For Use With (Equipment) | Hettich Angle Rotors |
| Vessels per Adapter | 1 |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur