Learn More
- Home
- Products
- Centrifuges
- Centrifuge Adapters
- Hettich Lab Technology™ Adapter
Hettich Lab Technology™ Adapter
Product Code. 11387660| Product Code. | Capacity | For Use With (Equipment) | Max. Tube Diameter | Max. Tube Length | unitSize |
|---|---|---|---|---|---|
| 11387660 | 250/290 mL | Carrier 1752 | 62 mm | 100 mm | Each |
| 10741543 | 4 x 9/15 mL | Carrier 1481, 1495 | 17 mm | 100 mm | Each |
| 10108821 | 50 mL | Carrier 1490 | 34 mm | 100 mm | Each |
| 10485012 | 450/650/750 mL | Carrier 4880 | 96 mm | 147 mm | Each |
| 10198381 | 100 mL | Carrier 1481, 1490, 1495 | 44 mm | 100 mm | Each |
| 10511083 | 50 mL | Rotor 1620A, 1720, 1792, 4794, 5615, 4246 | 29 mm | 115 mm | Each |
| 11464988 | 4 x 50 mL | Carrier 1752 | 29 mm | 115 mm | Each |
| 10272680 | 1.1 to 1.4 mL, 2.6 to 3.4 mL, 2.7 to 3 mL, 1.6 to 5 mL, 5 mL, 6 mL | Rotor 1137, 1139, 1015, 1418, 1613, 1615, 1721, EBA 200/200S | 13 mm | 82 mm | Each |
| 10557971 | 0.2/0.4 mL | Rotor 1612 | 6 mm | 45 mm | Pack of 6 |
| 10708944 | 20 x 2.7 to 3.0 mL, 4.5 to 5 mL, 5/6 mL | Carrier 5051, 5628 | 12 mm | 92 mm | Each |
| 11488259 | 9 x 1.5/2.0 mL | Carrier 1425 | 11 mm | 38 mm | Each |
| 10368962 | 17 x 15 mL | Carrier 5625-A | 17 mm | 100 mm | Each |
| 10450665 | 3 x 94/100 mL | Carrier 4780 | 65 mm | 115 mm | Each |
| 10767303 | 3 x 25/30 mL | Carrier 4795, 5645, 4266, 4570 | 26 mm | 100 mm | Each |
| 10198811 | 5 x 0.5/1.5/2.0 mL | Carrier 1481, 1490, 1495 | 11 mm | 38 mm | Each |
| 10274863 | 250/290 mL | Carrier 4890 | 62 mm | 137 mm | Each |
| 11435068 | 20 x 1.6 to 5/4 to 7 mL, 4 to 5.5/4.5 to 5 mL, 5/6 mL | Carrier 5625-A | 13 mm | 100 mm | Each |
| 12053924 | 9 x 1.5/2.0 mL | Carrier 1366, 1427 | 11 mm | 38 mm | Each |
| 10752354 | 12 x 1.6 to 5 mL, 2.6 to 3.4 mL, 4.9 mL, 5/6/7 mL | Carrier 5051, 5628 | 13 mm | 100 mm | Each |
| 11438199 | 100 mL | 100mL ASTM- Tube, Carrier 4275 | 58 mm | 161 mm | Each |
| 10046881 | 50 mL | 1 x 50mL Tubes | 34 mm | 100 mm | Each |
| 11424988 | 100 mL | Carrier 1752 | 44 mm | 100 mm | Each |
| 10629633 | 50 mL | Carrier 4880 | 29 mm | 115 mm | Each |
| 10301932 | 50 mL | Carrier 1481, 1490, 1495 | 29 mm | 115 mm | Each |
Product Code. 11387660
Brand: Hettich Lab Technology™ 1769
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
- Available in a variety of configurations
- Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
Specifications
| Type | Adapter |
| Capacity | 250/290 mL |
| Max. Tube Diameter | 62 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Carrier 1752 |
| No. per Pack | 1 Pack |