Learn More
Hettich Lab Technology™ Adapter
Product Code. 10752354
Click to view available options
Capacity:
0.2/0.4 mL
1.1 to 1.4 mL, 2.6 to 3.4 mL, 2.7 to 3 mL, 1.6 to 5 mL, 5 mL, 6 mL
100 mL
12 x 1.6 to 5 mL, 2.6 to 3.4 mL, 4.9 mL, 5/6/7 mL
17 x 15 mL
20 x 1.6 to 5/4 to 7 mL, 4 to 5.5/4.5 to 5 mL, 5/6 mL
20 x 2.7 to 3.0 mL, 4.5 to 5 mL, 5/6 mL
250/290 mL
3 x 25/30 mL
3 x 94/100 mL
4 x 50 mL
4 x 9/15 mL
450/650/750 mL
5 x 0.5/1.5/2.0 mL
50 mL
9 x 1.5/2.0 mL
For Use With (Equipment):
1 x 50mL Tubes
100mL ASTM- Tube, Carrier 4275
Carrier 1366, 1427
Carrier 1425
Carrier 1481, 1490, 1495
Carrier 1481, 1495
Carrier 1490
Carrier 1752
Carrier 4780
Carrier 4795, 5645, 4266, 4570
Carrier 4880
Carrier 4890
Carrier 5051, 5628
Carrier 5625-A
Rotor 1137, 1139, 1015, 1418, 1613, 1615, 1721, EBA 200/200S
Rotor 1612
Rotor 1620A, 1720, 1792, 4794, 5615, 4246
Max. Tube Diameter:
6 mm
11 mm
12 mm
13 mm
17 mm
26 mm
29 mm
34 mm
44 mm
58 mm
62 mm
65 mm
96 mm
Max. Tube Length:
38 mm
45 mm
82 mm
92 mm
100 mm
115 mm
137 mm
147 mm
161 mm
missing translation for 'unitSize'
Each
Pack of 6
Product Code. 10752354
missing translation for 'mfr': Hettich Lab Technology™ 5268
Hettich Lab Technology™ Adapter is designed to accommodate customized centrifuge workloads.
  • Available in a variety of configurations
  • Configurations capable of accommodating a host of customized centrifuge workloads
Order Info
dimForUseWithEquipment
Specifications
Type Adapter
Capacity 12 x 1.6 to 5 mL, 2.6 to 3.4 mL, 4.9 mL, 5/6/7 mL
Max. Tube Diameter 13 mm
Max. Tube Length 100 mm
For Use With (Equipment) Carrier 5051, 5628
No. per Pack 1 Each