Learn More
Hettich Lab Technology™ Adapter
Product Code. 10708944
Change view
Click to view available options
Capacity:
0.2/0.4 mL
1.1 to 1.4 mL, 2.6 to 3.4 mL, 2.7 to 3 mL, 1.6 to 5 mL, 5 mL, 6 mL
100 mL
12 x 1.6 to 5 mL, 2.6 to 3.4 mL, 4.9 mL, 5/6/7 mL
17 x 15 mL
20 x 1.6 to 5/4 to 7 mL, 4 to 5.5/4.5 to 5 mL, 5/6 mL
20 x 2.7 to 3.0 mL, 4.5 to 5 mL, 5/6 mL
250/290 mL
3 x 25/30 mL
3 x 94/100 mL
4 x 50 mL
4 x 9/15 mL
450/650/750 mL
5 x 0.5/1.5/2.0 mL
50 mL
9 x 1.5/2.0 mL
For Use With (Equipment):
1 x 50mL Tubes
100mL ASTM- Tube, Carrier 4275
Carrier 1366, 1427
Carrier 1425
Carrier 1481, 1490, 1495
Carrier 1481, 1495
Carrier 1490
Carrier 1752
Carrier 4780
Carrier 4795, 5645, 4266, 4570
Carrier 4880
Carrier 4890
Carrier 5051, 5628
Carrier 5625-A
Rotor 1137, 1139, 1015, 1418, 1613, 1615, 1721, EBA 200/200S
Rotor 1612
Rotor 1620A, 1720, 1792, 4794, 5615, 4246
Max. Tube Diameter:
6 mm
11 mm
12 mm
13 mm
17 mm
26 mm
29 mm
34 mm
44 mm
58 mm
62 mm
65 mm
96 mm
Max. Tube Length:
38 mm
45 mm
82 mm
92 mm
100 mm
115 mm
137 mm
147 mm
161 mm
Unit Size:
Each
Pack of 6
Product Code. Capacity For Use With (Equipment) Max. Tube Diameter Max. Tube Length unitSize
10708944 20 x 2.7 to 3.0 mL, 4.5 to 5 mL, 5/6 mL Carrier 5051, 5628 12 mm 92 mm Each
10741543 4 x 9/15 mL Carrier 1481, 1495 17 mm 100 mm Each
10108821 50 mL Carrier 1490 34 mm 100 mm Each
10485012 450/650/750 mL Carrier 4880 96 mm 147 mm Each
10198381 100 mL Carrier 1481, 1490, 1495 44 mm 100 mm Each
10511083 50 mL Rotor 1620A, 1720, 1792, 4794, 5615, 4246 29 mm 115 mm Each
11387660 250/290 mL Carrier 1752 62 mm 100 mm Each
11464988 4 x 50 mL Carrier 1752 29 mm 115 mm Each
10272680 1.1 to 1.4 mL, 2.6 to 3.4 mL, 2.7 to 3 mL, 1.6 to 5 mL, 5 mL, 6 mL Rotor 1137, 1139, 1015, 1418, 1613, 1615, 1721, EBA 200/200S 13 mm 82 mm Each
10557971 0.2/0.4 mL Rotor 1612 6 mm 45 mm Pack of 6
11488259 9 x 1.5/2.0 mL Carrier 1425 11 mm 38 mm Each
10368962 17 x 15 mL Carrier 5625-A 17 mm 100 mm Each
10450665 3 x 94/100 mL Carrier 4780 65 mm 115 mm Each
10767303 3 x 25/30 mL Carrier 4795, 5645, 4266, 4570 26 mm 100 mm Each
10198811 5 x 0.5/1.5/2.0 mL Carrier 1481, 1490, 1495 11 mm 38 mm Each
10274863 250/290 mL Carrier 4890 62 mm 137 mm Each
11435068 20 x 1.6 to 5/4 to 7 mL, 4 to 5.5/4.5 to 5 mL, 5/6 mL Carrier 5625-A 13 mm 100 mm Each
12053924 9 x 1.5/2.0 mL Carrier 1366, 1427 11 mm 38 mm Each
10752354 12 x 1.6 to 5 mL, 2.6 to 3.4 mL, 4.9 mL, 5/6/7 mL Carrier 5051, 5628 13 mm 100 mm Each
11438199 100 mL 100mL ASTM- Tube, Carrier 4275 58 mm 161 mm Each
10046881 50 mL 1 x 50mL Tubes 34 mm 100 mm Each
11424988 100 mL Carrier 1752 44 mm 100 mm Each
10629633 50 mL Carrier 4880 29 mm 115 mm Each
10301932 50 mL Carrier 1481, 1490, 1495 29 mm 115 mm Each
24 options
Product Code. 10708944
Brand: Hettich Lab Technology™ 5227
Hettich Lab Technology™ Adapter is designed to accommodate customized centrifuge workloads.
  • Available in a variety of configurations
  • Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
Type Adapter
Capacity 20 x 2.7 to 3.0 mL, 4.5 to 5 mL, 5/6 mL
Max. Tube Diameter 12 mm
Max. Tube Length 92 mm
For Use With (Equipment) Carrier 5051, 5628
No. per Pack 1 Pack