Learn More
Epredia™ Adamas Blades for Disposable Magnetic Blade Carriers
Shop All Epredia Products
Click to view available options
Length (Metric):
60 mm
Quantity:
10/Cs.
50/Cs.
Unit Size:
Pack of 10
Pack of 50
Specifications
Specifications
| Description | 60 x 18mm; 10/cs. |
| Product Type | Disposable, Magnetic |
| For Use With (Equipment) | Disposable Magnetic Blade Carrier |
| Quantity | 10/Cs. |
| Length (Metric) | 60 mm |
| Width (Metric) | 18 mm |