missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACVR1 (Human) Recombinant Protein
Human ACVR1 full-length ORF ( AAH33867, 1 a.a. - 509 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000090-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVDGVMILPVLIMIALPSPSMEDEKPKVNPKLYMCVCEGLSCGNEDHCEGQQCFSSLSINDGFHVYQKGCFQVYEQGKMTCKTPPSPGQAVECCQGDWCNRNITAQLPTKGKSFPGTQNFHLEVGLIILSVVFAVCLLACLLGVALRKFKRRNQERLNPRDVEYGTIEGLITTNVGDSTLADLLDHSCTSGSGSGLPFLVQRTVARQITLLECVGKGRYGEVWRGSWQGENVAVKIFSSRDEKSWFRETELYNTVMLRHENILGFIASDMTSRHSSTQLWLITHYHEMGSLYDYLQLTTLDTVSCLRIVLSIASGLAHLHIEIFGTQGKPAIAHRDLKSKNILVKKNGQCCIADLGLAVMHSQSTNQLDVGNNPRVGTKRYMAPEVLDETIQVDCFDSYKRVDIWAFGLVLWEVARRMVSNGIVEDYKPPFYDVVPNDPSFEDMRKVVCVDQQRPNIPNRWFSDPTLTSLAKLMKECWYQNPSARLTALRIKKTLTKIDNSLDKLKTDC
Specifications
AAH33867 | |
activin A receptor, type I | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACVR1 | |
GST |
90 | |
Wheat germ expression system | |
25 μg | |
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |