Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Click to view available options
Volume (Metric):
0.5-10 μL, 10-100 μL, 100-1000 μL
10, 100, 1000 μL
10, 50, 200 μL
10-100 μL, 100-1000 μL, 0.5-5.0 mL
100-1000 μL, 0.5-5.0, 1.0-10 mL
2, 10, 50 μL
2, 20, 200 μL
2-20 μL, 20-200 μL, 100-1000 μL
5-50 μL, 20-200 μL, 100-1000 μL
Unit Size:
1 set
Description
- Ergonomic shape.
- Lightweight.
- Uultra-soft plunger stroke.
- Smart and reliable volume adjustment:
- State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
- Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
- Digital display of set volume on the front of the instrument is easily visible during pipetting.
- Autoclavable.
Specifications
Specifications
| Angle | 360° |
| Certifications/Compliance | EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices |
| Autoclavable | Autoclavable |
| Product Line | Acura |
| Type | Micropipette Triopack |
| Volume (Metric) | 2, 10, 50 μL |
| Includes | 3 Acura™ micropipettes, manual, spike samples, Qualitips™ 2/10/50μL |
| Quantity | 3/Pk. |
| Warranty | 3 Years |