Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Description
- Air-displacement fixed-volume pipette.
- Exceptional, user-friendly ergonomics.
- Justip™ adjustable tip ejector fitting most tips.
- Shock, UV-light and autoclaving resistance.
- Ultra-soft plunger stroke.
- Available in a variety of volume models.
Specifications
Specifications
| Accuracy | <±0.7% |
| Certifications/Compliance | ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices). |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Model | 815 |
| Type | Micropipette |
| Volume (Metric) | 50 μL |
| Precision | <0.5% |
| Quantity | 1 Each |
| Warranty | 36 Months |