Learn More
Cole-Parmer™ Activated Carbon Filter Cartridge
Brand:  Cole-Parmer™ 01508-92
Additional Details : Weight : 0.41000kg
Product Code. 11739948
  • 292.00 NOK / Each
Estimated Shipment: 04-09-2024
Specifications
1 GPM
20 μm
1 Pack
254 mm
Activated Carbon
10 L
Filter Cartridge