Learn More
Cytiva Activated Calf Thymus DNA
Product Code. 10349874 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 Units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10349874 25 Units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10349874 Supplier Cytiva Supplier No. 27457501
  • ONLINE EXCLUSIVE PRICE
    3845.00 NOK
    3629.00 NOK / Pack of 1
    Save 216.00 NOK
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
High-quality activated calf thymus DNA
  • Treated with DNase I
For preparing substrate used in the study of DNA polymerases
TRUSTED_SUSTAINABILITY
Specifications
Format Liquid
Product Type Activated Calf Thymus DNA
Source Viral and eukaryote
Concentration 25 μg/mL
Quantity 25 Units
For Use With (Application) Study of DNA Polymerase