Learn More
Ansell ActivArmr™ 97-100 Oilfield Service Gloves with Ansell Grip™ Technology
Product Code. p-7215041 Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
Product Code. 16815270
Brand: Ansell 97100080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifically designed to meet the needs of production services workers working on or around oil and gas production sites. Ansell Grip™ Technology provides a sure grip in wet, oily environments for increased productivity and safety.
  • Enhanced grip and protection from oil penetration to enhance productivity
  • DuPont™ Aramid Fibers provide moderate cut resistance
  • Hi-Vis design is ideal for low light job sites and applications
  • Contact heat rated for protection against heated pipes and vessels
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE 0086, EN 407:2004, EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Material Nylon
Size 8
EN388 Standard 4242B
Glove Cuff Style Knit
Liner Material DuPont™ Aramid Fibers
Series 97-100
Yarn Gauge 15