missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ActivArmr™ 96-002 - Goatskin Leather Protectors

Product Code. 31114751 Shop All Ansell Products
Change view
Click to view available options
Model:
ActivArmr™ 96-002
ActivArmr™ 96-003
Size:
8
9
10
11
12
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Size
31114751 ActivArmr™ 96-002 9
31114752 ActivArmr™ 96-002 10
31114753 ActivArmr™ 96-002 11
31114754 ActivArmr™ 96-002 12
31114755 ActivArmr™ 96-003 8
31114756 ActivArmr™ 96-003 9
31114757 ActivArmr™ 96-003 10
31114758 ActivArmr™ 96-003 11
31114759 ActivArmr™ 96-003 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 31114751 Supplier Ansell™ Supplier No. 96002090
Add to basket
Add to basket
This item is not returnable. View return policy

Protective design for electrical insulated gloves that last longer with compliance to ASTM F696 and EN 388

  • Reinforced protection: ActivArmr™ 96-002 protectors are made from premium goatskin leather, enhancing hand safety when conducting electrical work with (or close to) energized components, while offering moderate abrasion, puncture and cut resistance
  • Improved durability: Designed to be worn over Class 0 & 00 electrical safety gloves, these leather protectors can significantly extend wear life
  • Assured effectiveness: ActivArmr™ 96-002 leather protectors meet ASTM D120 and EN 60903 standards, for personal protection you can truly depend on
  • Heightened dexterity: Their leather formulation also offers excellent dexterity, for genuinely easy-to-use electrical PPE
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ASTM F696∣Ecovadis Label∣EN 388
Construction Cut and Sewn
Coating Material Uncoated
Color White
Cut Resistance Rating Level B
Hand Specific
Latex Free Yes
Material Goatskin Leather
Product Type Industrial Gloves
Size 9
EN388 Standard 2111B
For Use With (Application) Electrical, Industrial, Automotive
Gender Unisex
Glove Cuff Color White
Glove Cuff Style Straight
Grip Design Textured
Liner Material Premium Goatskin
Lining Lined
Manufacturing/QMS Audit Standards ISO 9001:2015
Model ActivArmr™ 96-002
Packaging Type Carton
Length (English) 10 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.