Learn More
Ansell ActivArmr™ 27-905 Nitrile-Coated Gloves with Ansell Grip™ Technology
Product Code. 16875190 Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
Product Code. 16875190
Brand: Ansell 27905090
Only null left
Add to basket
Add to basket
New and improved nitrile coating is flexible and comfortable, while also providing mechanical protection. Ansell Grip™ Technology requires less force on slippery objects, reduces hand fatigue, and increases productivity.
  • Ideal for tough applications with a combination of three technologies: Ansell Grip™ Technology, Fortix™ Technology and Ripel™ Technology
  • Ansell's unique nitrile formulation is now more durable with patented Fortix™ Technology to offer resistance to abrasive and sharp materials
  • Ansell Grip™ Technology enhances grip in dry, wet, and oily environments
  • Full coating with Ripel™ Technology provides an outstanding barrier to grease and oil, without degrading like leather or cotton
  • Soft polycotton liner provides a cooler feeling to the hands
  • Safety cuff extends protection up the wrist
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Polycotton, Nitrile
Product Type Heavy-duty Nitrile Gloves
Size 9
EN388 Standard 4221C
For Use With (Application) Machinery, Equipment Operation, Oil Drilling, and Maintenance
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material Polycotton
Series 27-905