missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Antibody (CLIP/1407) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
5357.10 NOK
Specifications
| Antigen | ACTH |
|---|---|
| Clone | CLIP/1407 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry : 0.5-1ug/million cells, ELISA, Immunohistochemistry-Paraffin : 0.5-1ug/ml, Immunofluorescence : 1-2ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159387
|
Novus Biologicals
NBP2-47683-0.1MG |
0.1 mg |
5665.00 NOK 5357.10 NOK / 0.10mg Save 307.90 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ACTH | |
| 1.0 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Nutrient Sensing in the Brain | |
| PBS with No Preservative | |
| 5443 | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CLIP/1407 | |
| Flow Cytometry : 0.5-1ug/million cells, ELISA, Immunohistochemistry-Paraffin : 0.5-1ug/ml, Immunofluorescence : 1-2ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| POMC | |
| IgG | |
| Protein A or G purified | |
| ACTH (same as Corticotropin) is a 39 amino acid active peptide produced by the anterior pituitary. This MAb is specific to CLIP (aa25-39 of ACTH); does not react with Synacthen (aa1-24 of ACTH). POMC (pro-opiomelanocortin or corticotropin-lipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. POMC is cleaved into ten hormone chains named NPP, ACTH, alpha-MSH (Melanocyte Stimulating Hormone), beta-MSH, gamma-MSH, CLIP (corticotropin-like intermediary peptide), Lipotropin-beta, Lipotropin-gamma, beta-endorphin and Met-enkephalin. ACTH is also produced by cells of immune system (T-cells, B-cells, and macrophages) in response to stimuli associated with stress. Anti-ACTH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with ACTH-producing cells (corticotrophs). It also may react with other tumors (e.g. some small cell carcinomas of the lung) causing paraneoplastic syndromes by secreting ACTH. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title