missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17138-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| ACSS1 | |
| Polyclonal | |
| Unconjugated | |
| ACSS1 | |
| ACAS2L, AceCS2, AceCS2L, Acetate CoA ligase 2, Acetyl CoA synthetase 2, ACSS1, KIAA1846 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296259, 68738, 84532 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99NB1, Q9NUB1 | |
| Acss1 | |
| ACSS1 Fusion Protein Ag10896 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction