Learn More
Invitrogen™ ACSM4 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570980
Product Code. 15809393
  • 4230.00 NOK / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACSM4
Polyclonal
Unconjugated
ACSM4
ACSM4; acyl-CoA synthetase medium chain family member 4; acyl-CoA synthetase medium-chain family member 4; Acyl-coenzyme A synthetase ACSM4, mitochondrial; Olfactory specific medium-chain acyl CoA synthetase; Omacs; O-MACS
Rabbit
Affinity chromatography
RUO
341392
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P0C7M7
ACSM4
synthetic peptide directed towards the middle region of human ACSM4.