missing translation for 'onlineSavingsMsg'
Learn More

ACSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7207339 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855664 20 μL 20µL
16845664 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855664

Brand: Proteintech 161401AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA. Has some preference toward medium-chain substrates. Plays a role in adipocyte differentiation.

Specifications

Antigen ACSF2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACSF2
Gene Accession No. Q499N5, Q8VCW8, Q96CM8
Gene Alias ACSF2, ACSMW, AVYV493
Gene Symbols Acsf2
Host Species Rabbit
Immunogen ACSF2 Fusion Protein Ag9199
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 264895, 619561, 80221
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.