Learn More
Acrylic acid, tech. 90%, stab., Thermo Scientific™
Product Code. p-7059945
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
Product Code. Quantity unitSize
15492437 500 mL 500mL
15402447 2.5 L 2.5 L
2 options
Product Code. 15492437
Brand: Thermo Scientific Alfa Aesar 045779.AP
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-10-7
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O
Specifications
Melting Point ∼13°C
Density 1.051 g/mL
Boiling Point 138°C to 139°C
Flash Point 54°C (129°F)
Odor Pungent
Linear Formula H2C=CHCO2H
Refractive Index 1.4202
Quantity 500 mL
UN Number UN2218
Beilstein 635743
Merck Index 14,130
Solubility Information Completely miscible in water.
Formula Weight 72.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Acrylic acid, Stabilized