missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, ≥99%, MP Biomedicals™

Product Code. 11453071 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
11453071 100 g 100g
11463071 500 g 500g
11473071 1 kg 1kg
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11453071

Brand: MP Biomedicals 0215025680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Acrylamide, ≥99%, White Crystalline Powder

  • Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
  • It polymerizes in the presence of free radicals in aqueous solutions.
  • It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
  • Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym 2-Propenamide, Acrylic acid amide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Crystalline Powder
Density 1.127g/cm3 at 25°C
Color White
Quantity 100 g
Assay Percent Range ≥99%
Formula Weight 71.1
Chemical Name or Material Acrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.