Learn More
Cytiva (formerly Pall Lab) AcroPak™ 800 Capsules with Fluorodyne™ II Membrane
AcroPak 800 Capsules with Fluorodyne II (PVDF) membrane that is compatible with aqueous and many organic solvents.
Brand: Cytiva (formerly Pall Lab) 12471
Additional Details : Weight : 0.16500kg
Description
- Double layer sterilizing membrane enhances reliability
- Upstream air vent prevents vapor lock
- Manufactured without the use of adhesives to minimize extractables
- Pre-sterilized by gamma irradiation
Specifications
Hydrophilic PVDF | |
Sterile | |
Stepped Hose Barb |
0.2 μm | |
800 cm2 | |
1/Ea. |