Learn More
Cytiva AcroPak™ 300 Capsule with PTFE Membrane
Product Code. 17124221
Change view
Click to view available options
No. per Pack:
100/Pk.
3/Pk.
Unit Size:
Pack of 100
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
17124221 100/Pk. Pack of 100
17114221 3/Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17124221 Supplier Cytiva Supplier No. 12085
  • ONLINE EXCLUSIVE PRICE
    98079.07 NOK
    85597.96 NOK / Pack of 100
    Save 12481.11 NOK
    12% Off
Estimated Shipment: 15-10-2026
AcroPak 300 Capsule offers broad chemical compatibility with PTFE membrane and polypropylene housing.
  • High air flow rate venting applications
  • Offers broad chemical compatibility with PTFE membrane and polypropylene housing 100% integrity-tested to provide the high level of confidence you require
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 280 cm2
No. per Pack 100/Pk.
Outlet Style Stepped Hose Barb