Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ EKV Membrane
Product Code. 17124241
Click to view available options
Outlet Style:
1 to 1.5 in. Sanitary Flange
1/2 in. Hose Barb
Unit Size:
Each
Product Code. 17124241
Brand: Cytiva 12466
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 1500 cm2
Outlet Style 1 to 1.5 in. Sanitary Flange
Quantity 1/Ea.