Learn More
Cytiva AcroPak™ 1000 Capsules with Supor™ Membrane
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Specifications
| Material (Membrane) | Hydrophilic PES |
| Pore Size | 0.8/0.45 μm |
| Sterility | Sterile |
| Filtration Area | 1000 cm2 |
| Outlet Style | Stepped Hose Barb |
| Quantity | 1/Ea. |