Learn More
Thermo Scientific™ AcroMetrix SARS-CoV-2 Control (In-process, RUO)
Product Code. 16696382 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
1 set
Product Code. Quantity unitSize
16696382 Each 1 set
1 options
Product Code. 16696382
Brand: Thermo Scientific™ 954519
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO).
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO). This is a positive control kit containing two concentrations: a Low Positive and Ultra-Low Positive concentration to avoid false-negative results.
  • Full length genomic RNA from SARS-CoV-2
  • Kit contains one Low Positive control and one Ultra-low Positive control
  • Quantitated using Bio-Rad Droplet Digital™ PCR (ddPCR™)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C or lower
Description AcroMetrix Coronavirus 2019 (COVID-19) RNA Control (RUO)
Packaging One kit contains 2 vials per kit x 0.02mL per vial
Quantity Each
For Research Use Only.