Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Nylon Membrane and Male Slip Luer Outlet
Versatile filter for both aqueous and solvent-based sample filtration.
Brand: Cytiva (formerly Pall Lab) 4541
Additional Details : Weight : 1.02600kg
Description
- Excellent chemical compatibility with esters, bases, and alcohols
- Saves time because no prewetting is required
Specifications
Nylon | |
Non-sterile | |
<30 μL | |
1000/Pk. | |
55°C |
0.45 μm | |
1 cm2 | |
Polypropylene | |
Male Slip Luer Outlet | |
13 mm |