Learn More
Cytiva (formerly Pall Lab) Acro™ 37 TF Vent Device with PTFE Membrane
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.
Brand: Cytiva (formerly Pall Lab) 4464
3022.92 NOK valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 0.26000kg
Description
- PTFE membrane and polypropylene housing have broad chemical compatibility.
- This product is not approved for use in medical, clinical, surgical or other patient applications.
Specifications
PTFE | |
Non-sterile | |
24/Pk. |
0.2 μm | |
7.5 cm2 | |
37 mm |