Learn More
ACP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. p-7207163 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16894324 20 μL 20µL
16884324 150 μL 150µL
2 options
Product Code. 16894324
Brand: Proteintech 152361AP20UL
Rabbit Polyclonal Antibody
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
Antigen ACP2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACP2
Gene Accession No. P11117, P20611, P24638
Gene Alias acid phosphatase 2, lysosomal, ACP2, LAP, Lysosomal acid phosphatase
Gene Symbols ACP2
Host Species Rabbit
Immunogen ACP2 Fusion Protein Ag7356
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11432, 24162, 53
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG