missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11134-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.Specifications
| Aconitase 2 | |
| Polyclonal | |
| Unconjugated | |
| ACO2 | |
| ACO2, Aconitase, aconitase 2, aconitase 2, mitochondrial, ACONM, Citrate hydro lyase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11429, 50, 79250 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99798, Q99KI0, Q9ER34 | |
| ACO2 | |
| Aconitase 2 Fusion Protein Ag1590 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction