missing translation for 'onlineSavingsMsg'
Learn More

Acinus Antibody, Novus Biologicals™

Product Code. 18517730 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
Product Code. Quantity unitSize
18517730 0.025 mg 25µg
1 options
This item is not returnable. View return policy

Product Code. 18517730

Brand: Novus Biologicals NBP1768690.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acinus
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAD56724
Gene Alias Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107
Gene Symbols ACIN1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to amino acids near the C-terminus of the cleaved active peptide p17, which are identical to those of mouse Acinus. The immunogen is located within amino acids 1050 - 1100 of Acinus. Amino Acid Squence: TRTALHGVKWPQSNPK
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 22985
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.