Learn More
Acetylcholine Chloride Crystalline MP Biomedicals
Product Code. 11388279 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388279 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11388279 Supplier MP Biomedicals Supplier No. 0210001280
  • ONLINE EXCLUSIVE PRICE
    2475.00 NOK
    2353.18 NOK / 100g
    Save 121.82 NOK
    4% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Neurotransmitter with key applications in ophthalmology and biochemical research.
Specifications
CAS 60-31-1
Molecular Formula C7H16ClNO2
Quantity 100 g
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 181.66
PubChem CID 6060
ChEBI CHEBI:2417
Formula Weight 181.7
Percent Purity ∼99%
Physical Form Crystalline
Chemical Name or Material Acetylcholine Chloride