Learn More
Acetylacetone, Supelco™
Product Code. 15885340
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15885340 100 mL 100mL
15817740 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15885340 Supplier Supelco Supplier No. 1096000100
 Restricted Item
Estimated Shipment: 23-11-2026
For analysis EMSURE™
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-54-6
Grade Research
Molecular Formula CH3COCH2COCH3
Molecular Weight (g/mol) 100.12
Specifications
Melting Point −23°C (Lit.)
Density 0.975 g/mL at 25°C
Boiling Point 140.4°C
MDL Number MFCD00008787
Quantity 100 mL
Synonym ACAC, 2,4-Pentanedione, 2,4-Pentanedione