missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl CoA synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16087-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.Specifications
| Acetyl CoA synthetase | |
| Polyclonal | |
| Unconjugated | |
| ACSS2 | |
| ACAS2, AceCS, Acetate CoA ligase, Acetyl CoA synthetase, ACS, ACSA, ACSS2, Acyl activating enzyme | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311569, 55902, 60525 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NR19, Q9QXG4 | |
| Acss2 | |
| Acetyl CoA synthetase Fusion Protein Ag9121 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction