missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl CoA synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16087-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.Tekniske data
| Acetyl CoA synthetase | |
| Polyclonal | |
| Unconjugated | |
| ACSS2 | |
| ACAS2, AceCS, Acetate CoA ligase, Acetyl CoA synthetase, ACS, ACSA, ACSS2, Acyl activating enzyme | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311569, 55902, 60525 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NR19, Q9QXG4 | |
| Acss2 | |
| Acetyl CoA synthetase Fusion Protein Ag9121 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion