Learn More
Acetophenone, Sigma-Aldrich™
Specifications
Specifications
| Density | 1.03 g/cm3 at 20°C |
| MDL Number | MFCD00008724 |
| Quantity | 1 L |
| Synonym | Acetophenone, Methyl phenyl ketone, 1-Phenylethanone, Phenylethanone, ACP, Acetophenone |