missing translation for 'onlineSavingsMsg'
Learn More

Acetonyltriphenylphosphonium chloride, 99%

Product Code. 11417696
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11417696 100 g 100g
11407696 25 g 25g
11427696 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11417696

Brand: Thermo Scientific Alfa Aesar A14691.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1235-21-8 | C21H20ClOP | 354.81 g/mol

Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

Solubility
Soluble in hot water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1235-21-8
Molecular Formula C21H20ClOP
Molecular Weight (g/mol) 354.81
MDL Number MFCD00011813
InChI Key XAMZZEBAJZJERT-UHFFFAOYSA-M
Synonym acetonyltriphenylphosphonium chloride, 2-oxopropyl triphenylphosphonium chloride, 2-oxopropyltriphenylphosphonium chloride, phosphonium, acetonyltriphenyl-, chloride, acetonyl triphenylphosphonium chloride, phosphonium, 2-oxopropyl triphenyl-, chloride, acetonyltriphenylphosphoniumchloride, 2-oxopropyl triphenylphosphanium chloride, 1-triphenylphosphino acetone, chloride, acmc-1c01z
PubChem CID 196994
IUPAC Name 2-oxopropyl(triphenyl)phosphanium;chloride
SMILES [Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C to 248°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in hot water.
Formula Weight 354.82
Percent Purity 99%
Chemical Name or Material Acetonyltriphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.