Learn More
Acetonitrile - D3, deuteration degree min. 99% for NMR spectroscopy MagniSolv™, MilliporeSigma™
Product Code. 15874310
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Septum Ampule
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15874310 10 mL Septum Ampule 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15874310 Supplier Sigma-Aldrich Supplier No. 1029040010
 Restricted Item
Estimated Shipment: 21-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For NMR Spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Odor Ether-like
Packaging Septum Ampule
Vapor Pressure 97 hPa at 20°C (68°F)
Viscosity 0.39mPa.s (20°C)
Quantity 10 mL
CAS 2206-26-0
Assay Percent Range 99% min.
Molecular Formula C2H3N
MDL Number MFCD00001881
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES [2H]C([2H])([2H])C#N
IUPAC Name (²H₃)acetonitrile
Molecular Weight (g/mol) 44.07
PubChem CID 123151
Formula Weight 44.07
Chemical Name or Material Acetonitrile - D3