Learn More
Acetamiprid, Fluka™
Product Code. 16292892 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16292892
Brand: Honeywell-Fluka R1607100MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetamiprid
Packaging Glass Bottle
CAS 135410-20-7
Color White
Melting Point 98°C
Boiling Point Decomposes below the boiling point.
Synonym N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine
Industry Type Environmental Analysis
Molecular Weight (g/mol) 222.67
Quantity 100 mg
Grade Analytical Standard