missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, Sigma-Aldrich™

Product Code. 15876760
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876760 100 g 100g
15831821 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15876760 Supplier Sigma Aldrich Supplier No. 8223430100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-35-5
Grade Research
Molecular Weight (g/mol) 59.07

Specifications

Melting Point 78°C to 81°C
Density 1.159 g/cm3 at 20°C
Boiling Point 221°C to 222°C/1013 hPa
MDL Number MFCD00008023
Quantity 100 g
Synonym Acetamide, Acetic acid amide, Acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.